Please login to your online account to display your discounted pricing

Ammonium sulfate, Reagent grade, 99%, MP Biomedicals™

$59.44 - $59.44

Chemical Identifiers

Chemical Name or Material Ammonium sulfate
CAS 7783-20-2
Purity Grade Reagent Grade
Physical Form Crystalline Powder
Assay Percent Range 0.99
Molecular Formula (NH4)2SO4
Formula Weight 132.1
Synonym Sulfuric acid diammonium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN10477290

 
mp biomedicals
10477290
500g Each for $59.44
Description & Specifications

Specifications

Formula Weight 132.1
Purity Grade Reagent Grade
Physical Form Crystalline Powder
Color White
Quantity 500g
Density 1.77g/cm3 at 20°C
CAS 7783-20-2
Chemical Name or Material Ammonium sulfate
Assay Percent Range 0.99
Molecular Formula (NH4)2SO4
Synonym Sulfuric acid diammonium salt