missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium sulfate, Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1063909

 View more versions of this product

Catalog No. AA1063909



Specifications

Specifications

Ammonium sulfate
(metals basis)
(NH4)2SO4
14,555
BFNBIHQBYMNNAN-UHFFFAOYSA-N
diazanium;sulfate
6097028
132.14
99.999%
1.769g/mL
~280°C (decomposition)
7783-20-2
H8N2O4S
MFCD00003391
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
[NH4+].[NH4+].[O-]S(=O)(=O)[O-]
132.134
CHEBI:62946
Crystalline
Puratronic™
White
10g

Chemical Identifiers

7783-20-2
132.134
BFNBIHQBYMNNAN-UHFFFAOYSA-N
6097028
diazanium;sulfate
H8N2O4S
MFCD00003391
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
CHEBI:62946
[NH4+].[NH4+].[O-]S(=O)(=O)[O-]
Safety and Handling

Safety and Handling

RTECSNumber : BS4500000

TSCA : TSCA

Documents