missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Sulfate (Laboratory), Fisher Chemical™

$342.93 - $3,888.93

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.134
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym actamaster, ammonium sulfate, ammonium sulfate 2:1, ammonium sulphate, ammoniumsulfate, diammonium sulfate, diammonium sulphate, mascagnite, sulfuric acid diammonium salt, sulfuric acid, diammonium salt
PubChem CID 6097028
ChEBI CHEBI:62946
IUPAC Name diazanium;sulfate
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
A701-3
SDS View Documents
Fisher Chemical
FLA7013
3kg Glass Bottle
Each for $342.93
Case of 4 Each for $1,371.72
 
A701-50
SDS Accessories View Documents
Fisher Chemical
FLA70150
50kg Fiber Drum
Each for $3,888.93
 
Description

Description

Specifications

Chemical Identifiers

7783-20-2
132.134
BFNBIHQBYMNNAN-UHFFFAOYSA-N
6097028
diazanium;sulfate
H8N2O4S
MFCD00003391
actamaster, ammonium sulfate, ammonium sulfate 2:1, ammonium sulphate, ammoniumsulfate, diammonium sulfate, diammonium sulphate, mascagnite, sulfuric acid diammonium salt, sulfuric acid, diammonium salt
CHEBI:62946
[NH4+].[NH4+].[O-]S(=O)(=O)[O-]

Specifications

7783-20-2
MFCD00003391
BFNBIHQBYMNNAN-UHFFFAOYSA-N
diazanium;sulfate
6097028
132.14
Laboratory
Pass Test
Colorless
5 to 6
Ammonium Sulfate
H8N2O4S
actamaster, ammonium sulfate, ammonium sulfate 2:1, ammonium sulphate, ammoniumsulfate, diammonium sulfate, diammonium sulphate, mascagnite, sulfuric acid diammonium salt, sulfuric acid, diammonium salt
[NH4+].[NH4+].[O-]S(=O)(=O)[O-]
132.134
CHEBI:62946
Solid
1.77g/mL
Glass Bottle
280°C
3kg
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. . .

NFPA

Health:1
Flammability:0
Instability:0

SDS