Please login to your online account to display your discounted pricing

Ammonium Sulfate, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$318.21 - $318.21

Chemical Identifiers

Chemical Name or Material Ammonium Sulfate
CAS 7783-20-2
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula (NH4)2SO4
Formula Weight 132.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002327

 
avantor performance
079205
Poly Bottle 2.5kg Each for $318.21
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 132.14
Physical Form Granular
Chemical Name or Material Ammonium Sulfate
Specific Gravity 1L = 1.77kg
Packaging Poly Bottle
Molecular Formula (NH4)2SO4
CAS 7783-20-2
Quantity 2.5kg