Please login to your online account to display your discounted pricing

Ammonium Sulfate, For Radium, Certified, 10.0%(w/v) ±0.5%(w/v), LabChem™

$95.26 - $95.26

Chemical Identifiers

Chemical Name or Material Ammonium Sulfate
CAS 7732-18-5
CAS Max % 90.51
CAS 7783-20-2
CAS Max % 9.49
Name Note For Radium
Purity Grade Certified Grade
Linear Formula (NH4)2SO4
Formula Weight 132.13
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC113001

 
lab chem
LC113001
500mL Each for $95.26
Description & Specifications

Specifications

Formula Weight 132.13
Purity Grade Certified Grade
Assay 10.0%± 0.5%(w/v)
Solubility Soluble in water
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Packaging Poly Bottle
Chemical Name or Material Ammonium Sulfate
CAS Max % 9.49
CAS 7783-20-2
Traceability to NIST Traceable to NIST
Density 1.05g/mL
Physical Form Liquid
CAS 7732-18-5
Linear Formula (NH4)2SO4
Viscosity 1.1cSt
CAS Max % 90.51
Quantity 500mL
Decomposition Information Gaseous Ammonia; Sulfur compounds
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Storage Information Room Temperature
Specific Gravity 1.05g/mL
Color Clear, colorless
Identification Passes Test
Name Note For Radium
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.