Please login to your online account to display your discounted pricing

Ammonium Sulfate, Fisher BioReagents


Quantity 1kg
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLBP212R1

 View more versions of this product

Catalog No. BP212R-1

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.5%
Formula Weight 132.14
Melting Point 280°C
Quantity 1kg
Protease Protease free
Physical Form Solid
Color Colorless
pH 5 to 6
Viscosity [NULL]
Merck Index 15, 552
Heavy Metals (as Pb) 5ppm max.
Ignition Residue 0.005% max.
Iron (Fe) 5ppm max.
Packaging Poly Bottle
Molecular Formula H8N2O4S
Linear Formula (NH4)2SO4
ChemAlert Storage Symbol Gray
Arsenic (As) 0.5ppm max.
Synonym Diammonium Sulfate
Health Hazard 2 CAUTION!
Absorbance 0.025 max. at 280nm
Chemical Name or Material Ammonium Sulfate
Identification Pass Test
Nitrate 0.001 max.
CAS 7783-20-2
Health Hazard 3 Emergency Overview
May cause skin, eye, and respiratory tract irritation. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. . .

NFPA

Health:1
Flammability:0
Instability:0
Recommended Storage RT

Specifications

Chemical Name or Material Ammonium Sulfate
Assay ≥99.5%
Molecular Formula H8N2O4S
Linear Formula (NH4)2SO4
Formula Weight 132.14
Synonym Diammonium Sulfate
Accessories
 

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. . .

NFPA

Health:1
Flammability:0
Instability:0

CAUTION!

RT

SDS & Safety Documents