missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Sulfate, BiotechGrade, 99%, Spectrum™ Chemical

A1249, 7783-20-2, (NH4)2SO4

$316.74 - $3,384.88

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
IUPAC Name sulfuric acid diamine
SMILES N.N.OS(O)(=O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18600653
View Documents Promotion Details
Spectrum Chemical Mfg Cor
A1249500GM
500 g Poly Bottle
Each for $316.74
 
18600654
View Documents Promotion Details
Spectrum Chemical Mfg Cor
A124925KG
2.5 kg Poly Bottle
Each for $959.12
 
18600655
View Documents Promotion Details
Spectrum Chemical Mfg Cor
A124912KGBL
12 kg Poly Pail
Each for $3,384.88
 
Description

Description

Spectrum™ Chemical Ammonium Sulfate, BiotechGrade is an inorganic salt that is very soluble in water It is a widely used reagent in molecular biology and chromatography for the precipitation and fractionation of proteins, crystallization of proteins and protein-nucleic acid complexes. Spectrum offers highly pure reagents suitable for biochemical research and analysis.
Specifications

Chemical Identifiers

7783-20-2
132.13
sulfuric acid diamine
H8N2O4S
BFNBIHQBYMNNAN-UHFFFAOYSA-N
N.N.OS(O)(=O)=O

Specifications

7783-20-2
500 g
H8N2O4S
N.N.OS(O)(=O)=O
132.13
Biotech
0.005%
Poly Bottle
5.0-6.0
White, crystallized
5 ppm
100%
None detected
BFNBIHQBYMNNAN-UHFFFAOYSA-N
sulfuric acid diamine
99%
5 ppm
0.001%
5 ppm
0.005%
5 ppm
5 ppm
SDS
Product Certifications