missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium Sulfate, BiotechGrade, 99%, Spectrum™ Chemical
A1249, 7783-20-2, (NH4)2SO4
Supplier: Spectrum Chemical Mfg Cor A124925KG
Description
Spectrum™ Chemical Ammonium Sulfate, BiotechGrade is an inorganic salt that is very soluble in water It is a widely used reagent in molecular biology and chromatography for the precipitation and fractionation of proteins, crystallization of proteins and protein-nucleic acid complexes. Spectrum offers highly pure reagents suitable for biochemical research and analysis.Specifications
7783-20-2 | |
5.0-6.0 | |
H8N2O4S | |
BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
sulfuric acid diamine | |
99% | |
Poly Bottle | |
White, crystallized |
100% | |
2.5 kg | |
MFCD00003391 | |
N.N.OS(O)(=O)=O | |
132.13 | |
Biotech | |
0.005% |
Chemical Identifiers
7783-20-2 | |
132.13 | |
BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
N.N.OS(O)(=O)=O |
H8N2O4S | |
MFCD00003391 | |
sulfuric acid diamine |