Please login to your online account to display your discounted pricing

Ammonium Sulfate, ACS Grade, ≥ 99.0%, LabChem™

$59.03 - $59.03

Chemical Identifiers

Chemical Name or Material Ammonium Sulfate
CAS 7783-20-2
CAS Max % 100
Purity Grade ACS Grade
Linear Formula (NH4)2SO4
Formula Weight 132.13
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC112981

 
lab chem
LC112981
500g Each for $59.03
Description & Specifications

Specifications

Assay ≥99.0%
Formula Weight 132.13
Chemical Name or Material Ammonium Sulfate
EINECS Number 231-984-1
Identification Passes Test
Packaging Poly Bottle
Purity Grade ACS Grade
Physical Form Granules
Solubility Soluble in water
Specific Gravity 1.77g/mL
Storage Information Room Temperature
Linear Formula (NH4)2SO4
CAS 7783-20-2
Decomposition Information Gaseous Ammonia; Sulfur compounds
Color White
Melting Point 280°C
Quantity 500g
CAS Max % 100
Health Hazard 2 GHS H Statement
Harmful to aquatic life.
Health Hazard 3 GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.
Density 1.77g/mL
Safety and Handling

GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful to aquatic life.