missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Sulfate ACS AR Granular, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 7783-20-2
Physical Form Granular
Molecular Formula (NH4)2SO4
Formula Weight 132.14
Chemical Name or Material Ammonium Sulfate
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK3512500 Avantor Performance
351212
POLYSTORMOR 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7783-20-2
(NH4)2SO4
Ammonium Sulfate
Granular
132.14
ACS Grade

Specifications

7783-20-2
(NH4)2SO4
POLYSTORMOR
Ammonium Sulfate
Granular
132.14
500g
ACS Grade