Please login to your online account to display your discounted pricing

Ammonium Sulfate ACS AR Granular, Macron Fine Chemicals™


macron

Quantity 125g
Packaging POLYSTORMOR

Manufacturer: avantor performance  351202

 View more versions of this product

Catalog No. MK3512125

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Packaging POLYSTORMOR
Quantity 125g
Chemical Name or Material Ammonium Sulfate
CAS 7783-20-2
Purity Grade ACS
Physical Form Granular
Molecular Formula (NH4)2SO4
Formula Weight 132.14g/mol

Specifications

Chemical Name or Material Ammonium Sulfate
CAS 7783-20-2
Purity Grade ACS
Physical Form Granular
Molecular Formula (NH4)2SO4
Formula Weight 132.14g/mol