Please login to your online account to display your discounted pricing

Ammonium sulfate, ACS, 99.0%, Alfa Aesar™

In galvanizing steel, manufacturing of ammonia alum and sulfuric acid

$52.43 - $52.43

Chemical Identifiers

Chemical Name or Material Ammonium sulfate
CAS 7783-20-2
Purity Grade ACS Grade
Molecular Formula H8N2O4S
Linear Formula (NH4)2SO4
Formula Weight 132.14
MDL Number MFCD00003391
Merck Index 14,555
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1156636

  SDS
alfa aesar
1156636
Each for $52.43
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 132.14
Quantity 500g
Chemical Name or Material Ammonium sulfate
TSCA TSCA
Physical Form Granular
Assay ≥99.0%
Molecular Formula H8N2O4S
Linear Formula (NH4)2SO4
MDL Number MFCD00003391
CAS 7783-20-2
Merck Index 14,555
Color White
Melting Point ≈280°C (Dec.)
Solubility Information Soluble in water. Insoluble in alcohol and acetone
Density 1.769
RTECS Number BS4500000