Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium sulfate, ≥99%, Molecular Biology Grade

$126.22 - $126.22

Chemical Identifiers

Chemical Name or Material Ammonium sulfate
CAS 7783-20-2
Purity Grade Molecular Biology Grade
Linear Formula (NH4)2SO4
Formula Weight 132.14
MDL Number MFCD00003391
Merck Index 14,555
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ64180A1

 
alfa aesar™
J64180A1
1kg Each for $126.22
Description & Specifications

Specifications

Formula Weight 132.14
Chemical Name or Material Ammonium sulfate
Assay ≥99%
MDL Number MFCD00003391
CAS 7783-20-2
Melting Point ≈280°C (dec.)
TSCA TSCA
Purity Grade Molecular Biology Grade
Linear Formula (NH4)2SO4
Color White
Quantity 1kg
Density 1.769g/mL
RTECS Number BS4500000
Physical Form Crystalline
Merck Index 14,555