missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium sulfate, 99%, Acros Organics™

Manufacturer:  Acros Organics AC447135000

 View more versions of this product

Catalog No. AC447135000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Ammonium sulfate
H8N2O4S
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
Solubility in water: 754g/L. Other solubilities: insoluble in alcohol and acetone
[NH4+].[NH4+].[O-]S(=O)(=O)[O-]
132.134
CHEBI:62946
99%
1ppm max.
5ppm max.
5ppm max.
2ppm max.
5ppm max.
1ppm max.
4.5 to 5.5 (5% aq. soln.)
7783-20-2
(NH4)2SO4
1ppm max.
BFNBIHQBYMNNAN-UHFFFAOYSA-N
diazanium;sulfate
6097028
132.14
Biochemical
10ppm max.
1ppm max.
0.005% max.
10ppm max.
Solubility in water: 754g/L. Other solubilities: insoluble in alcohol and acetone
280.0°C
500g

Chemical Identifiers

7783-20-2
132.134
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
CHEBI:62946
[NH4+].[NH4+].[O-]S(=O)(=O)[O-]
H8N2O4S
BFNBIHQBYMNNAN-UHFFFAOYSA-N
6097028
diazanium;sulfate
Documents