missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium sulfate, 99.95% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 8936336

 View more versions of this product

Catalog No. AA8936336



Specifications

Specifications

Ammonium sulfate
99.95% (metals basis)
MFCD00003391
BFNBIHQBYMNNAN-UHFFFAOYSA-N
diazanium;sulfate
6097028
132.14
500g
7783-20-2
H8N2O4S
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
[NH4+].[NH4+].[O-]S(=O)(=O)[O-]
132.134
CHEBI:62946
Crystalline

Chemical Identifiers

7783-20-2
132.134
BFNBIHQBYMNNAN-UHFFFAOYSA-N
6097028
diazanium;sulfate
H8N2O4S
MFCD00003391
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
CHEBI:62946
[NH4+].[NH4+].[O-]S(=O)(=O)[O-]
Safety and Handling

Safety and Handling

EINECSNumber : 231-984-1

SDS Documents