Please login to your online account to display your discounted pricing

Ammonium sulfate, 99.5%, for analysis, ACROS Organics™


Quantity 5kg
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC205870050

 View more versions of this product

Catalog No. AC205870050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 132.14
Quantity 5kg
Purity Grade For analysis
Density 1.76
CAS 7783-20-2
Chemical Name or Material Ammonium sulfate
MDL Number MFCD00003391
Packaging Plastic bottle
Linear Formula (NH4)2SO4
Assay Percent Range 99.5%
Molecular Formula H8N2O4S
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.

Specifications

Chemical Name or Material Ammonium sulfate
CAS 7783-20-2
Purity Grade For analysis
Assay Percent Range 99.5%
Molecular Formula H8N2O4S
Linear Formula (NH4)2SO4
Formula Weight 132.14
MDL Number MFCD00003391
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents