Learn More
Ammonium sulfate, 99.5%, for analysis, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 205872500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ammonium sulfate | |
7783-20-2 | |
99.5% | |
(NH4)2SO4 | |
15,552 | |
Solubility in water: 754g/L. Other solubilities: insoluble in alcohol and acetone | |
N.N.OS(O)(=O)=O | |
132.13 | |
CHEBI:62946 | |
99.5% | |
Plastic bottle | |
1.76 |
280.0°C | |
250 g | |
H8N2O4S | |
MFCD00003391 | |
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate | |
BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
diazanium;sulfate | |
6097028 | |
132.14 | |
Analytical | |
0.01% max. (as SO4) |
Chemical Identifiers
7783-20-2 | |
132.13 | |
BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
6097028 | |
N.N.OS(O)(=O)=O |
H8N2O4S | |
MFCD00003391 | |
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate | |
CHEBI:62946 |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 231-984-1
RUO – Research Use Only