Please login to your online account to display your discounted pricing

Ammonium Sulfate, 10% (w/v), Ricca Chemical

$86.73 - $86.73

Chemical Identifiers

Chemical Name or Material Ammonium Sulfate
CAS 7732-18-5
CAS Min % 89.0
CAS Max % 93.0
CAS 7783-20-2
CAS Min % 9.0
CAS Max % 11.0
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

694-216

 
ricca chemical company
RI694.216
Poly natural 500mL Each for $86.73

694-232

 
ricca chemical company
RI694.232
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 500mL
Concentration or Composition Notes 10% (w/v)
CAS Min % 89.0
CAS Max % 93.0
CAS Min % 9.0
CAS Max % 11.0
Chemical Name or Material Ammonium Sulfate
CAS 7732-18-5
CAS 7783-20-2
Packaging Poly natural