Please login to your online account to display your discounted pricing

Ammonium Sulfamate, 99%, (Crystalline), MP Biomedicals


Quantity 1kg

Manufacturer: mp biomedicals  0219385991

 View more versions of this product

Catalog No. ICN19385991

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1kg
Chemical Name or Material Ammonium Sulfamate
CAS 7773-06-0
Physical Form Crystalline
Assay Percent Range 99%
Molecular Formula NH4SO3NH2
Formula Weight 114.1g/mol
Recommended Storage Room Temperature

Specifications

Chemical Name or Material Ammonium Sulfamate
CAS 7773-06-0
Physical Form Crystalline
Assay Percent Range 99%
Molecular Formula NH4SO3NH2
Formula Weight 114.1g/mol
Accessories

Room Temperature