Please login to your online account to display your discounted pricing

Ammonium Sulfamate, 99%, (Crystalline), MP Biomedicals

$343.88 - $550.29

Chemical Identifiers

Chemical Name or Material Ammonium Sulfamate
CAS 7773-06-0
Physical Form Crystalline
Assay Percent Range 99%
Molecular Formula NH4SO3NH2
Formula Weight 114.1g/mol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19385990

 
mp biomedicals
0219385990
500g Each for $343.88

ICN19385991

 
mp biomedicals
0219385991
1kg Each for $550.29
Description & Specifications

Specifications

Quantity 500g
Chemical Name or Material Ammonium Sulfamate
CAS 7773-06-0
Physical Form Crystalline
Assay Percent Range 99%
Molecular Formula NH4SO3NH2
Formula Weight 114.1g/mol
Recommended Storage Room Temperature
Safety and Handling

Room Temperature