missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium sulfamate, 98% min., ACS, Alfa Aesar™

Manufacturer:  Alfa Aesar 14534A3

 View more versions of this product

Catalog No. AA14534A3



Specifications

Specifications

Ammonium sulfamate
7773-06-0
H6N2O3S
ammonium sulfamate, ammonium sulphamate, ammate, ikurin, ammonium amidosulfonate, feliderm k, ammate x, sulfamic acid, monoammonium salt, ammonium sulfamidate, ammonium sulphamidate
[NH4+].NS(=O)(=O)[O-]
114.119
CHEBI:81950
Crystalline
For ACS analysis
98% min.
MFCD00011429
GEHMBYLTCISYNY-UHFFFAOYSA-N
azanium;sulfamate
24482
114.12
2kg

Chemical Identifiers

7773-06-0
114.119
GEHMBYLTCISYNY-UHFFFAOYSA-N
24482
azanium;sulfamate
H6N2O3S
MFCD00011429
ammonium sulfamate, ammonium sulphamate, ammate, ikurin, ammonium amidosulfonate, feliderm k, ammate x, sulfamic acid, monoammonium salt, ammonium sulfamidate, ammonium sulphamidate
CHEBI:81950
[NH4+].NS(=O)(=O)[O-]
Safety and Handling

Safety and Handling

EINECSNumber : 231-871-7

Documents