Please login to your online account to display your discounted pricing

Ammonium Sulfamate, ≥98.0%, (ACS Reagent Grade), MP Biomedicals


Quantity 100g

Manufacturer: mp biomedicals  15249980

 View more versions of this product

Catalog No. ICN15249980

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Formula Weight 114.1
Quantity 100g
Chemical Name or Material Ammonium Sulfamate
Assay Percent Range ≥98.0%
Molecular Formula NH4OSO2NH2
CAS 7773-06-0
Recommended Storage Room Temperature

Specifications

Chemical Name or Material Ammonium Sulfamate
CAS 7773-06-0
Purity Grade ACS Reagent Grade
Assay Percent Range ≥98.0%
Molecular Formula NH4OSO2NH2
Formula Weight 114.1
Accessories

Room Temperature