Please login to your online account to display your discounted pricing

Ammonium Sulfamate, ≥98.0%, (ACS Reagent Grade), MP Biomedicals

$43.44 - $130.37

Chemical Identifiers

Chemical Name or Material Ammonium Sulfamate
CAS 7773-06-0
Purity Grade ACS Reagent Grade
Assay Percent Range ≥98.0%
Molecular Formula NH4OSO2NH2
Formula Weight 114.1
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN15249980

 
mp biomedicals
15249980
100g Each for $43.44

ICN15249990

 
mp biomedicals
15249990
500g Each for $130.37
Description & Specifications

Specifications

Purity Grade ACS Reagent Grade
Formula Weight 114.1
Quantity 100g
Chemical Name or Material Ammonium Sulfamate
Assay Percent Range ≥98.0%
Molecular Formula NH4OSO2NH2
CAS 7773-06-0
Recommended Storage Room Temperature
Safety and Handling

Room Temperature