missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium phosphomolybdate hydrate, Reagent Grade

Manufacturer:  Alfa Aesar™ 1798436

 View more versions of this product

Catalog No. AA1798436



Specifications

Specifications

Ammonium phosphomolybdate hydrate
H80Mo12N3O41P-6
MFCD00036312
BIQVMZMYTFVCJZ-UHFFFAOYSA-H
azane;molybdenum;trihydroxide;phosphate;tetratriacontahydrate
131664298
Powder
3.15g/mL
500g
54723-94-3
(NH4)3PO4·12MoO3·xH2O (x≈3)
ammonium phosphomolybdate hydrate
N.N.N.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[OH-].[OH-].[OH-].[O-]P(=O)([O-])[O-].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo]
1960.994
1876.35 (Anhydrous)
Reagent
Yellow

Chemical Identifiers

54723-94-3
1960.994
BIQVMZMYTFVCJZ-UHFFFAOYSA-H
131664298
N.N.N.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[OH-].[OH-].[OH-].[O-]P(=O)([O-])[O-].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo]
H80Mo12N3O41P-6
MFCD00036312
ammonium phosphomolybdate hydrate
azane;molybdenum;trihydroxide;phosphate;tetratriacontahydrate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents