missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Phosphate Monobasic, TraceSELECT™, ≥99.9999% (metals basis), Honeywell™ Fluka™

$295.60 - $1,022.34

Chemical Identifiers

CAS 7722-76-1
Molecular Formula H6NO4P
Molecular Weight (g/mol) 115.025
MDL Number MFCD00003396
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
Synonym ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
PubChem CID 24402
ChEBI CHEBI:62982
IUPAC Name azanium;dihydrogen phosphate
SMILES [NH4+].OP(=O)(O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60044613 View Documents Honeywell Chemicals
09726100G
100g Poly Bottle Each for $1,022.34
 
60044614 View Documents Honeywell Chemicals
0972625G
25g Poly Bottle Each for $295.60
 
Specifications

Chemical Identifiers

7722-76-1
115.025
LFVGISIMTYGQHF-UHFFFAOYSA-N
24402
azanium;dihydrogen phosphate
H6NO4P
MFCD00003396
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
CHEBI:62982
[NH4+].OP(=O)(O)[O-]

Specifications

7722-76-1
NH4H2PO4
NONH for all modes of transport
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
[NH4+].OP(=O)(O)[O-]
115.025
CHEBI:62982
≥99.9999% (metals basis)
1.803g/cu.cm
<0.00001 hPa at 20°C
Colorless
3.8 to 4.4 (at 50g/L 25°C)
Ammonium Phosphate Monobasic
H6NO4P
MFCD00003396
Powder/Crystals
LFVGISIMTYGQHF-UHFFFAOYSA-N
azanium;dihydrogen phosphate
24402
115.03g/mol
TraceSELECT™
Poly Bottle
197°C
190°C
100g
Safety and Handling

Safety and Handling

EINECSNumber : 231-764-5