Please login to your online account to display your discounted pricing

Ammonium Phosphate, Monobasic, 99.0% min., ACS, EMD Millipore

$1,973.37 - $1,973.37

Chemical Identifiers

Chemical Name or Material Ammonium Phosphate, Monobasic
Name Note EMSURE™, For Analysis
CAS 7722-76-1
Purity Grade ACS/EP/Reagent Grade
Assay Percent Range 99.0% min.
Molecular Formula H6NO4P
Linear Formula (NH4)H2PO4
Formula Weight 115.2
Synonym Ammonium Biphosphate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MAX13507

 
emd millipore corporation
AX13507
12kg Each for $1,973.37
Description & Specifications

Specifications

Purity Grade ACS/EP/Reagent Grade
Formula Weight 115.2
Quantity 12kg
Bulk Density 800 to 1100kg/m3
Packaging Poly Pail
Synonym Ammonium Biphosphate
Melting Point 19°C
Density 1.80g/cm3 (20°C)
Chemical Name or Material Ammonium Phosphate, Monobasic
Name Note EMSURE™, For Analysis
Assay Percent Range 99.0% min.
Molecular Formula H6NO4P
Linear Formula (NH4)H2PO4
CAS 7722-76-1
Vapor Pressure 0.066 hPa (125°C)
Recommended Storage Store without restrictions
Safety and Handling

Store without restrictions