Please login to your online account to display your discounted pricing

Ammonium Phosphate, Dibasic, ACS Grade, ≥ 98.0%, LabChem™

$120.60 - $120.60

Chemical Identifiers

Chemical Name or Material Ammonium Phosphate
CAS 7783-28-0
CAS Max % 100
Name Note Dibasic
Purity Grade ACS Grade
Linear Formula (NH4)2HPO4
Formula Weight 132.06
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC112851

 
lab chem
LC112851
500g Each for $120.60
Description & Specifications

Specifications

Assay ≥98.0%
Formula Weight 132.06
Chemical Name or Material Ammonium Phosphate
EINECS Number 231-987-8
Identification Passes Test
Name Note Dibasic
Packaging Poly Bottle
Purity Grade ACS Grade
Physical Form Crystals
Solubility Soluble in water
Specific Gravity 1.619g/mL
Storage Information Room Temperature
Linear Formula (NH4)2HPO4
CAS 7783-28-0
Decomposition Information Gaseous Ammonia; Phosphorus oxides
Color White
Melting Point 155°C
Quantity 500g
CAS Max % 100
Health Hazard 2 GHS H Statement
Product is not hazardous.
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Density 1.619g/mL
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Product is not hazardous.