missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium phosphate, dibasic, 99%, for biochemistry, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 447142500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ammonium phosphate, dibasic | |
250 g | |
H9N2O4P | |
MFCD00010891 | |
diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate | |
MNNHAPBLZZVQHP-UHFFFAOYSA-N | |
diazanium;hydrogen phosphate | |
24540 | |
132.06 | |
Biochemical |
7783-28-0 | |
99% | |
(NH4)2HPO4 | |
15,54 | |
(1 M in water) Clear colorless | |
N.N.OP(O)(O)=O | |
132.06 | |
CHEBI:63051 | |
99% | |
Plastic bottle |
Chemical Identifiers
7783-28-0 | |
132.06 | |
MNNHAPBLZZVQHP-UHFFFAOYSA-N | |
24540 | |
N.N.OP(O)(O)=O |
H9N2O4P | |
MFCD00010891 | |
diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate | |
CHEBI:63051 |
Safety and Handling
Warning
EINECSNumber : 231-987-8
RUO – Research Use Only