Please login to your online account to display your discounted pricing

Ammonium phosphate, dibasic, 99+%, for analysis, ACROS Organics™


Quantity 250g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  AC201822500

 View more versions of this product

Catalog No. AC201822500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Formula Weight 132.06
Physical Form Adhering Crystals
Quantity 250g
Color White
Merck Index 15, 540
EINECS Number 231-987-8
TSCA TSCA
Density 1.61
Purity Grade For analysis
pH 7.9 to 8.3 (5% soln. at 20°C)
Chloride 5ppm max.
Sulfate 200ppm max.
Iron (Fe) 10ppm max.
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Ammonium phosphate, dibasic
MDL Number MFCD00010891
Molecular Formula H9N2O4P
Name Note p.a.
CAS 7783-28-0
Assay Percent Range 99+%
Linear Formula (NH4)2HPO4
Sodium (Na) 100ppm max.
Packaging Plastic bottle
Synonym DAP
Nitrate 100ppm max.
Heavy Metals (as Pb) 10ppm max.
Potassium (K) 100ppm max.
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

Specifications

Chemical Name or Material Ammonium phosphate, dibasic
Name Note p.a.
CAS 7783-28-0
Purity Grade For analysis
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula H9N2O4P
Linear Formula (NH4)2HPO4
Formula Weight 132.06
MDL Number MFCD00010891
Synonym DAP
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents