Please login to your online account to display your discounted pricing

Ammonium persulfate, Reagent grade, For ACS analysis, ≥98%, MP Biomedicals™


Quantity 100g

Manufacturer: mp biomedicals  19385880

 View more versions of this product

Catalog No. ICN19385880

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Solid
Formula Weight 228.2
Purity Grade ACS Reagent Grade
Color White
Quantity 100g
Density 950 to 1050kg/m3 (bulk density)
Name Note For ACS analysis
Molecular Formula (NH4)2S2O8
CAS 7727-54-0
Chemical Name or Material Ammonium Persulfate
Storage Note 1 Store dessicated.
Synonym Ammonium peroxydisulfate
Assay Percent Range ≥98%
Recommended Storage Store at room temperature.

Specifications

Chemical Name or Material Ammonium Persulfate
Name Note For ACS analysis
CAS 7727-54-0
Purity Grade ACS Reagent Grade
Physical Form Crystalline Solid
Assay Percent Range ≥98%
Molecular Formula (NH4)2S2O8
Formula Weight 228.2
Synonym Ammonium peroxydisulfate
Accessories

Store at room temperature.

Store dessicated.