missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium persulfate, Reagent grade, For ACS analysis, ≥98%, MP Biomedicals™

$63.18 - $63.18

Chemical Identifiers

CAS 7727-54-0
Physical Form Crystalline Solid
Assay Percent Range ≥98%
Molecular Formula (NH4)2S2O8
Formula Weight 228.2
Synonym Ammonium peroxydisulfate
Chemical Name or Material Ammonium Persulfate, For ACS analysis
Grade ACS Reagent Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19385880 Mp Biomedicals
19385880
100g Each for $63.18
Specifications Description & Specifications

Chemical Identifiers

7727-54-0
≥98%
228.2
Ammonium Persulfate, For ACS analysis
Crystalline Solid
(NH4)2S2O8
Ammonium peroxydisulfate
ACS Reagent Grade

Specifications

7727-54-0
≥98%
228.2
950 to 1050kg/m3 (bulk density)
100g
ACS Reagent Grade
Crystalline Solid
(NH4)2S2O8
Ammonium peroxydisulfate
White
Ammonium Persulfate, For ACS analysis