Please login to your online account to display your discounted pricing

Ammonium Persulfate, OmniPur™ Calbiochem™, EMD Millipore

$24.93 - $24.93

Chemical Identifiers

Assay Percent Range ≥98%
Formula Weight 228.2
Boiling Point 120°C
CAS 7727-54-0
Chemical Name or Material Ammonium Persulfate
Density 1.98g/cm3 at 25°C
Molecular Formula (NH4)2S2O8
Solubility Information Soluble in water
Recommended Storage Store in accordance with local regulations. Store in original container, protected from direct sunlight. Separate from reducing agents and combustible materials. Keep container tightly closed and sealed until ready for use.
Synonym Ammonium Peroxydisulfate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

230025GM

 
emd millipore corp
230025GM
Each for $24.93
Description & Specifications

Specifications

Boiling Point 120°C
Formula Weight 228.2
Quantity 25g
Synonym Ammonium Peroxydisulfate
Density 1.98g/cm3 at 25°C
CAS 7727-54-0
Molecular Formula (NH4)2S2O8
Recommended Storage Store in accordance with local regulations. Store in original container, protected from direct sunlight. Separate from reducing agents and combustible materials. Keep container tightly closed and sealed until ready for use.
Chemical Name or Material Ammonium Persulfate
Odor Odorless
Assay Percent Range ≥98%
Purity Molecular biology grade
UN Number UN1444
Solubility Information Soluble in water
DOT Information DOT Class: 5.1, Packaging Group: III

Initiator for acrylamide polymerization

Safety and Handling

Store in accordance with local regulations. Store in original container, protected from direct sunlight. Separate from reducing agents and combustible materials. Keep container tightly closed and sealed until ready for use.