Please login to your online account to display your discounted pricing

Ammonium Persulfate, MP Biomedicals™


Quantity 100g

Manufacturer: mp biomedicals  0219055680

 View more versions of this product

Catalog No. ICN19055680

  • / Each
 

Description & Specifications

Chemical Identifiers

Density 950 to 1050kg/m3 (bulk density)
Physical State Solid
Quantity 100g
Chemical Name or Material Ammonium Persulfate
CAS 7727-54-0
Physical Form White crystalline solid
Molecular Formula (NH4)2S2O8
Formula Weight 228.2g/mol
Synonym Ammonium peroxydisulfate
Storage Note 1 Store dessicated.
Recommended Storage Store at room temperature.

Specifications

Chemical Name or Material Ammonium Persulfate
CAS 7727-54-0
Physical Form White crystalline solid
Molecular Formula (NH4)2S2O8
Formula Weight 228.2g/mol
Synonym Ammonium peroxydisulfate
Accessories

Store at room temperature.

Store dessicated.