Please login to your online account to display your discounted pricing

Ammonium Persulfate, MP Biomedicals™

$110.96 - $110.96

Chemical Identifiers

Chemical Name or Material Ammonium Persulfate
CAS 7727-54-0
Physical Form Crystalline Solid
Molecular Formula (NH4)2S2O8
Formula Weight 228.2
Synonym Ammonium peroxydisulfate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19055691

 
mp biomedicals
19055691
1kg Each for $110.96
Description & Specifications

Specifications

Formula Weight 228.2
Physical Form Crystalline Solid
Color White
Quantity 1kg
Density 950 to 1050kg/m3 (bulk density)
Physical State Solid
Chemical Name or Material Ammonium Persulfate
Molecular Formula (NH4)2S2O8
CAS 7727-54-0
Synonym Ammonium peroxydisulfate
Storage Note 1 Store dessicated.
Recommended Storage Store at room temperature.