Please login to your online account to display your discounted pricing

Ammonium Persulfate, MP Biomedicals™


Quantity 1kg

Manufacturer: mp biomedicals  19055691

 View more versions of this product

Catalog No. ICN19055691

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 228.2
Physical Form Crystalline Solid
Color White
Quantity 1kg
Density 950 to 1050kg/m3 (bulk density)
Physical State Solid
Chemical Name or Material Ammonium Persulfate
Molecular Formula (NH4)2S2O8
Recommended Storage Store at room temperature.
Synonym Ammonium peroxydisulfate
CAS 7727-54-0
Storage Note 1 Store dessicated.

Specifications

Chemical Name or Material Ammonium Persulfate
CAS 7727-54-0
Physical Form Crystalline Solid
Molecular Formula (NH4)2S2O8
Formula Weight 228.2
Synonym Ammonium peroxydisulfate
Accessories

Store at room temperature.

Store dessicated.