Please login to your online account to display your discounted pricing

Ammonium Persulfate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$621.28 - $2,409.07

Chemical Identifiers

Chemical Name or Material Ammonium Persulfate
CAS 7727-54-0
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula (NH4)2S2O8
Formula Weight 228.2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002314

 
avantor performance
076205
Poly Bottle 2.5kg Case of 4 Each for $2,409.07

02002315

 
avantor performance
076201
Poly Bottle 500g Case of 4 Each for $621.28
Description & Specifications

Specifications

Physical Form Crystals
Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 228.2
Chemical Name or Material Ammonium Persulfate
CAS 7727-54-0
Molecular Formula (NH4)2S2O8
Quantity 2.5kg
Packaging Poly Bottle
Specific Gravity 1L = 1.98kg