Please login to your online account to display your discounted pricing

Ammonium Persulfate (Colorless-to-White Crystals/Electrophoresis), Fisher BioReagents


Quantity 100g
Packaging Glass Bottle

Manufacturer: fisher scientific company  BP179100

 View more versions of this product

Catalog No. BP179-100

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Formula Weight 228.19
Physical Form Powder/Solid
Melting Point 120°C
Purity Grade Electrophoresis Grade
Quantity 100g
DOT Information DOT Class 5.1, : Oxidizer
pH 3.2
Density 1.980g/cm³
Decomposition Information >120°C
Color White
Merck Index 15, 539
Heavy Metals (as Pb) 0.005% max.
Insoluble Matter 0.005% max.
Packaging Glass Bottle
Health Hazard 2 DANGER!
Chemical Name or Material Ammonium Persulfate
CAS 7727-54-0
ChemAlert Storage Symbol Yellow
Linear Formula (NH4)2S2O8
Additional Information Vapor Pressure: Negligible
Synonym APS
Molecular Formula H8N2O8S2
Manganese (Mn) 0.5ppm max.
Ignition Residue 0.05% max.
Identification Pass Test
Iron (Fe) 0.001% max.
Assay Percent Range ≥98.0 %
Titratable Free Acid 0.04mEq/g max.
Chloride and Chlorate 0.001% max.
Health Hazard 3 Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Causes eye, skin, and respiratory tract irritation. May cause allergic respiratory and skin reaction. Use personal protective equipment. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:2
Flammability:1
Instability:2
Recommended Storage RT

Specifications

Chemical Name or Material Ammonium Persulfate
CAS 7727-54-0
Purity Grade Electrophoresis Grade
Physical Form Powder/Solid
Assay ≥98.0%
Assay Percent Range ≥98.0 %
Molecular Formula H8N2O8S2
Linear Formula (NH4)2S2O8
Formula Weight 228.19
Synonym APS
Accessories
 

Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Causes eye, skin, and respiratory tract irritation. May cause allergic respiratory and skin reaction. Use personal protective equipment. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:2
Flammability:1
Instability:2

DANGER!

RT

SDS & Safety Documents