Learn More

Ammonium Persulfate (Colorless-to-White Crystals/Electrophoresis), Fisher BioReagents™
CAS: 7727-54-0 | H8N2O8S2 | 228.19 g/mol
$62.31 - $62.77
Chemical Identifiers
CAS | 7727-54-0 |
---|---|
Molecular Formula | H8N2O8S2 |
Molecular Weight (g/mol) | 228.19 |
MDL Number | MFCD00003390 |
InChI Key | ROOXNKNUYICQNP-UHFFFAOYSA-N |
Synonym | ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french |
PubChem CID | 62648 |
IUPAC Name | diazanium;sulfonatooxy sulfate |
SMILES | [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
BP179-25
|
Fisher BioReagents
BP17925 |
25 g | Glass Bottle |
Each for $62.31
Case of 6 Each for $373.86
|
|
||||
BP179-100
|
Fisher BioReagents
BP179100 |
100 g | Glass Bottle |
Each for $62.77
Case of 6 Each for $376.62
|
|
||||
Description
Ammonium Persulfate (Colorless-to-White Crystals/Electrophoresis)
Electrophoresis-grade Ammonium Persulfate is an initiator of acrylamide polymerization for the preparation of polyacrylamide electrophoresis gels.Chemical Identifiers
7727-54-0 | |
228.19 | |
ROOXNKNUYICQNP-UHFFFAOYSA-N | |
62648 | |
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O |
H8N2O8S2 | |
MFCD00003390 | |
ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french | |
diazanium;sulfonatooxy sulfate |
Specifications
120°C | |
White | |
Powder/Solid | |
≥98.0 % | |
(NH4)2S2O8 | |
15, 539 | |
ROOXNKNUYICQNP-UHFFFAOYSA-N | |
diazanium;sulfonatooxy sulfate | |
62648 | |
≥98.0% | |
Pass Test | |
Negligible | |
1.980g/cm³ |
7727-54-0 | |
3.2 | |
25 g | |
H8N2O8S2 | |
MFCD00003390 | |
ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french | |
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O | |
228.19 | |
228.19 | |
Electrophoresis | |
Glass Bottle | |
>120°C | |
Ammonium Persulfate |
Safety and Handling

DANGER!
Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Causes eye, skin, and respiratory tract irritation. May cause allergic respiratory and skin reaction. Use personal protective equipment. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.
NFPA
Health:2
Flammability:1
Instability:2
DOTInformation : DOT Class 5.1, : Oxidizer
Recommended Storage : RT