missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Persulfate, ACS, ≥98%, Ultrapure, Thermo Scientific™

High speed initiator used to catalyze acrylamide polymerization.

Manufacturer:  Thermo Fisher Scientific J76322A1

 View more versions of this product

Catalog No. AAJ76322A1



Specifications

Specifications

Ammonium Persulfate
H8N2O8S2
UN1444
ROOXNKNUYICQNP-UHFFFAOYSA-N
diazanium;sulfonatooxy sulfate
62648
Crystalline Powder
ACS
1.982
1kg
7727-54-0
MFCD00003390
ammonium peroxydisulfate
[NH4+].[NH4+].[O-]S(=O)(=O)OOS(=O)(=O)[O-]
228.19
228.19
≥98%
Plastic bottle
120°C (decomposition)

Chemical Identifiers

7727-54-0
228.19
ROOXNKNUYICQNP-UHFFFAOYSA-N
62648
[NH4+].[NH4+].[O-]S(=O)(=O)OOS(=O)(=O)[O-]
H8N2O8S2
MFCD00003390
ammonium peroxydisulfate
diazanium;sulfonatooxy sulfate
Safety and Handling

Safety and Handling

TSCA : Y

Documents