missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Ammonium Persulfate, ACS, ≥98%, Ultrapure, Thermo Scientific™

High speed initiator used to catalyze acrylamide polymerization.

$30.22 - $121.59

Chemical Identifiers

CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
MDL Number MFCD00003390
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
Synonym ammonium peroxydisulfate, ammonium persulfate, ammoniumpersulfate, ccris 1430, diammonium peroxodisulphate, diammonium peroxydisulfate, diammonium peroxydisulphate, diammonium persulfate, persulfate d'ammonium french, unii-22qf6l357f
PubChem CID 62648
IUPAC Name diazanium;sulfonatooxy sulfate
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS(=O)(=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AAJ7632209
View Documents
Thermo Scientific™
J7632209
10g Plastic bottle Each for $30.22
 
AAJ7632222
View Documents
Thermo Scientific™
J7632222
100g Plastic bottle Each for $54.27
 
AAJ76322A1
View Documents
Thermo Scientific™
J76322A1
1kg Plastic bottle Each for $121.59
 
Specifications

Chemical Identifiers

7727-54-0
228.19
ROOXNKNUYICQNP-UHFFFAOYSA-N
62648
[NH4+].[NH4+].[O-]S(=O)(=O)OOS(=O)(=O)[O-]
H8N2O8S2
MFCD00003390
ammonium peroxydisulfate, ammonium persulfate, ammoniumpersulfate, ccris 1430, diammonium peroxodisulphate, diammonium peroxydisulfate, diammonium peroxydisulphate, diammonium persulfate, persulfate d'ammonium french, unii-22qf6l357f
diazanium;sulfonatooxy sulfate

Specifications

7727-54-0
MFCD00003390
ammonium peroxydisulfate, ammonium persulfate, ammoniumpersulfate, ccris 1430, diammonium peroxodisulphate, diammonium peroxydisulfate, diammonium peroxydisulphate, diammonium persulfate, persulfate d'ammonium french, unii-22qf6l357f
[NH4+].[NH4+].[O-]S(=O)(=O)OOS(=O)(=O)[O-]
228.19
228.19
≥98%
1.982
1.982
10g
H8N2O8S2
UN1444
ROOXNKNUYICQNP-UHFFFAOYSA-N
diazanium;sulfonatooxy sulfate
62648
Crystalline Powder
ACS
Plastic bottle
120°C (decomposition)
AmmoniumPersulfate
Safety and Handling

Safety and Handling

P210-P220-P221-P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P333+P313-P342+P311-P362-P370+P378q-P501c

H272-H302-H315-H317-H319-H334-H335

DOTInformation : Hazard Class: 5.1; Packaging Group: III

EINECSNumber : 231-786-5

RTECSNumber : SE0350000

TSCA : Yes

Recommended Storage : Ambient temperatures