missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Ammonium Persulfate, ACS, ≥98%, Ultrapure, Thermo Scientific™

High speed initiator used to catalyze acrylamide polymerization.

Manufacturer:  Thermo Scientific™ J7632209

 View more versions of this product

Catalog No. AAJ7632209



Specifications

Specifications

Ammonium Persulfate
H8N2O8S2
UN1444
ROOXNKNUYICQNP-UHFFFAOYSA-N
diazanium;sulfonatooxy sulfate
62648
Crystalline Powder
ACS
Plastic bottle
120°C (decomposition)
7727-54-0
MFCD00003390
ammonium peroxydisulfate, ammonium persulfate, ammoniumpersulfate, ccris 1430, diammonium peroxodisulphate, diammonium peroxydisulfate, diammonium peroxydisulphate, diammonium persulfate, persulfate d'ammonium french, unii-22qf6l357f
[NH4+].[NH4+].[O-]S(=O)(=O)OOS(=O)(=O)[O-]
228.19
228.19
≥98%
1.982
1.982
10g

Chemical Identifiers

7727-54-0
228.19
ROOXNKNUYICQNP-UHFFFAOYSA-N
62648
[NH4+].[NH4+].[O-]S(=O)(=O)OOS(=O)(=O)[O-]
H8N2O8S2
MFCD00003390
ammonium peroxydisulfate, ammonium persulfate, ammoniumpersulfate, ccris 1430, diammonium peroxodisulphate, diammonium peroxydisulfate, diammonium peroxydisulphate, diammonium persulfate, persulfate d'ammonium french, unii-22qf6l357f
diazanium;sulfonatooxy sulfate
Safety and Handling

Safety and Handling

P210-P220-P221-P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P333+P313-P342+P311-P362-P370+P378q-P501c

H272-H302-H315-H317-H319-H334-H335

DOTInformation : Hazard Class: 5.1; Packaging Group: III

EINECSNumber : 231-786-5

RTECSNumber : SE0350000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents