Please login to your online account to display your discounted pricing

Ammonium persulfate, >98%, Molecular biology reagent grade, MP Biomedicals™


Quantity 100g

Manufacturer: mp biomedicals  0219398880

 View more versions of this product

Catalog No. ICN19398880

  • / Each
 

Description & Specifications

Chemical Identifiers

Density 951 to 1050 kg/m3 (bulk density)
Quantity 100g
Chemical Name or Material Ammonium persulfate
CAS 7727-54-0
Purity Grade >98%, Molecular biology reagent grade
Physical Form White crystalline powder
Molecular Formula (NH4)2S2O8
Formula Weight 228.18g/mol
Synonym Ammonium peroxydisulfate

Specifications

Chemical Name or Material Ammonium persulfate
CAS 7727-54-0
Purity Grade >98%, Molecular biology reagent grade
Physical Form White crystalline powder
Molecular Formula (NH4)2S2O8
Formula Weight 228.18g/mol
Synonym Ammonium peroxydisulfate