Learn More
Ammonium persulfate, 98%, extra pure, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 201530050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Oxidation reagentSpecifications
Ammonium persulfate | |
5kg | |
H8N2O8S2 | |
MFCD00003390 | |
15, 539 | |
Solubility in water: 582g/L (20°C),may decompose | |
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O | |
228.19 | |
228.19 | |
Extra Pure | |
1.98 |
7727-54-0 | |
98% | |
(NH4)2S2O8 | |
01,952; 02,348; 03,238; 05,15; 06,20; 12,33 | |
ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french | |
ROOXNKNUYICQNP-UHFFFAOYSA-N | |
diazanium;sulfonatooxy sulfate | |
62648 | |
98% | |
Plastic Bottle | |
1.9800g/mL |
Safety and Handling
GHS H Statement
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Take any precaution to avoid mixing with combustibles.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs.
GHS Signal Word: Danger
EINECSNumber : 231-786-5
RUO â Research Use Only