missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Fluka™ Ammonium Persulfate, ≥98.0%, ACS Reagent, Honeywell™ Fluka™

ACS Reagent,≥98.0%

$610.58 - $1,961.23

Chemical Identifiers

CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
MDL Number MFCD00003390
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
Synonym ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
PubChem CID 62648
IUPAC Name diazanium;sulfonatooxy sulfate
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS(=O)(=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60045665 View Documents Honeywell Fluka™
24861425KG
2.5kg Poly Bottle Each for $610.58
 
60045666 View Documents Honeywell Fluka™
2486144X25KG
4 x 2.5kg Poly Bottle Case of 4 Each for $1,961.23
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7727-54-0
228.19
ROOXNKNUYICQNP-UHFFFAOYSA-N
62648
[NH4+].[NH4+].[O-]S(=O)(=O)OOS(=O)(=O)[O-]
H8N2O8S2
MFCD00003390
ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
diazanium;sulfonatooxy sulfate

Specifications

7727-54-0
(NH4)2S2O8
UN1444
ROOXNKNUYICQNP-UHFFFAOYSA-N
diazanium;sulfonatooxy sulfate
62648
≥98.0%
1.98g/cm3
≤0.05%
Solid
120°C
Ammonium Persulfate
H8N2O8S2
MFCD00003390
ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
[NH4+].[NH4+].[O-]S(=O)(=O)OOS(=O)(=O)[O-]
228.19
228.2g/mol
ACS Reagent
Poly Bottle
7.9 (vs air)
Colorless
2.5kg
Safety and Handling

Safety and Handling

P220-P261-P280-P305 + P351 + P338-P342 + P311

H272-H302-H315-H317-H319-H334-H335

DOTInformation : 5.1/PGIII

EINECSNumber : 231-786-5

RTECSNumber : SE0350000