Learn More
Ammonium perrhenate(VII), 99+% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01141018
Description
Ammonium perrhenate(VII) is used to synthesis the rehenium(III) organohydrazides which is used as radiopharmaceuticals and also used in the manufacturing of rhenium metal and its additives in superalloys. It is used as catalyst for the synthesis of platinum-rhenium catalysts which used in gasoline.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ammonium perrhenate(VII) | |
13598-65-7 | |
Powder | |
(metals basis) | |
NH4ReO4 | |
UN1479 | |
Soluble in water. | |
N.O[Re](=O)(=O)=O | |
268.24 | |
268.24 | |
3.97 g/mL |
(decomposition) | |
White | |
50 g | |
H4NO4Re | |
MFCD00012377 | |
azanium oxido trioxo rhenium, ammonium tetraoxorhenate vii, acmc-20alsh, nh4reo4, rhenate reo41-, ammonium, t-4, ammonium tetraoxorhenate 1-, rhenate reo41-, ammonium 1:1 , t-4, ammonium perrhenate, nh4 reo4, rhenate reo41-, ammonium, beta-4, ammonium perrhenate vii , puratronic metals basis | |
XPSXHXWHJAIQJR-UHFFFAOYSA-M | |
azanium;oxido(trioxo)rhenium | |
3084163 | |
≥99% |
Chemical Identifiers
13598-65-7 | |
268.24 | |
XPSXHXWHJAIQJR-UHFFFAOYSA-M | |
3084163 |
H4NO4Re | |
MFCD00012377 | |
azanium oxido trioxo rhenium, ammonium tetraoxorhenate vii, acmc-20alsh, nh4reo4, rhenate reo41-, ammonium, t-4, ammonium tetraoxorhenate 1-, rhenate reo41-, ammonium 1:1 , t-4, ammonium perrhenate, nh4 reo4, rhenate reo41-, ammonium, beta-4, ammonium perrhenate vii , puratronic metals basis | |
N.O[Re](=O)(=O)=O |
Safety and Handling
GHS H Statement
H272
May intensify fire
oxidizer.
GHS P Statement
P221-P210-P220-P280-P370+P378a-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
EINECSNumber : 237-075-6
RTECSNumber : VI0710000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only