missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium perrhenate(VII), 99+% (metals basis)

Manufacturer:  Alfa Aesar™ 1141004

 View more versions of this product

Catalog No. AA1141004



Specifications

Specifications

Ammonium perrhenate(VII)
(metals basis)
NH4ReO4
UN1479
HOJYZCWLNWENHS-UHFFFAOYSA-O
azanium;oxido(trioxo)rhenium
3084163
Powder
3.97g/mL
2g
13598-65-7
H4NO4Re
MFCD00012377
acmc-20alsh
[NH4+].[O-][Re](=O)(=O)=O
268.242
268.24
≥99%
White

Chemical Identifiers

13598-65-7
268.242
HOJYZCWLNWENHS-UHFFFAOYSA-O
3084163
[NH4+].[O-][Re](=O)(=O)=O
H4NO4Re
MFCD00012377
acmc-20alsh
azanium;oxido(trioxo)rhenium
Safety and Handling

Safety and Handling

GHS H Statement
H272
May intensify fire
oxidizer.

GHS P Statement
P221-P210-P220-P280-P370+P378a-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.

RTECSNumber : VI0710000

TSCA : TSCA

Documents