missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium peroxydisulfate, Electrophoresis Grade

Manufacturer:  Alfa Aesar™ J6185622

 View more versions of this product

Catalog No. AAJ6185622



Specifications

Specifications

7727-54-0
(NH4)2S2O8
UN1444
ammonium peroxydisulfate, ammonium persulfate, ammoniumpersulfate, ccris 1430, diammonium peroxodisulphate, diammonium peroxydisulfate, diammonium peroxydisulphate, diammonium persulfate, persulfate d'ammonium french, unii-22qf6l357f
[NH4+].[NH4+].[O-]S(=O)(=O)OOS(=O)(=O)[O-]
228.19
228.2
Electrophoresis
Odorless
Moisture sensitive
120°C (decomposition)
None
H8N2O8S2
MFCD00003390
14,541
ROOXNKNUYICQNP-UHFFFAOYSA-N
diazanium;sulfonatooxy sulfate
62648
Powder
1.982
1.5
White
100g

Chemical Identifiers

7727-54-0
228.19
ROOXNKNUYICQNP-UHFFFAOYSA-N
62648
[NH4+].[NH4+].[O-]S(=O)(=O)OOS(=O)(=O)[O-]
H8N2O8S2
MFCD00003390
ammonium peroxydisulfate, ammonium persulfate, ammoniumpersulfate, ccris 1430, diammonium peroxodisulphate, diammonium peroxydisulfate, diammonium peroxydisulphate, diammonium persulfate, persulfate d'ammonium french, unii-22qf6l357f
diazanium;sulfonatooxy sulfate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H301-H334-H315-H319-H317-H335
May intensify fire; oxidizer.
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

P210-P220-P221-P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P333+P313-P342+P311-P362-P370+P378q-P501c

H272-H302-H315-H317-H319-H334-H335

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: AMMONIUM PERSULFATE

EINECSNumber : 231-786-5

RTECSNumber : SE0350000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents