missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium peroxydisulfate, ACS, 98.0% min, Alfa Aesar™

Manufacturer:  Alfa Aesar 54106A1

 View more versions of this product

Catalog No. AA54106A1



Specifications

Specifications

Ammonium peroxydisulfate
98.0% min
MFCD00003390
ROOXNKNUYICQNP-UHFFFAOYSA-N
diazanium;sulfonatooxy sulfate
62648
Lumps
1.982 g/mL
7727-54-0
H8N2O8S2
ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
[NH4+].[NH4+].[O-]S(=O)(=O)OOS(=O)(=O)[O-]
228.19
228.19
ACS
1kg

Chemical Identifiers

7727-54-0
228.19
ROOXNKNUYICQNP-UHFFFAOYSA-N
62648
[NH4+].[NH4+].[O-]S(=O)(=O)OOS(=O)(=O)[O-]
H8N2O8S2
MFCD00003390
ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
diazanium;sulfonatooxy sulfate
Safety and Handling

Safety and Handling

EINECSNumber : 231-786-5

SDS Documents