Please login to your online account to display your discounted pricing

Ammonium Peroxodisulfate, EMD Millipore


For analysis EMSURE™ ACS, Reag. Ph Eur

Quantity 50kg
Packaging Fibre carton

Manufacturer: emd millipore corporation  1012019050

 View more versions of this product

Catalog No. M1012019050

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 228.19
Quantity 50kg
Health Hazard 1 H272: May intensify fire; oxidiser.
RTECS Number SE0350000
Health Hazard 3 H315: Causes skin irritation.
Density 1.98g/cm3 at 20°C
Health Hazard 2 H302: Harmful if swallowed.
Chemical Name or Material Ammonium peroxodisulfate
DOT Information DOT Class: 5.1, Packaging Group: III
CAS 7727-54-0
EINECS Number 231-786-5
Packaging Fibre carton
UN Number UN1444
Synonym Ammonium persulfate; Peroxidisulfuric acid diammonium salt
Recommended Storage Store at 15° to 25°C (59° to 77°F), Tightly closed. Dry. Away from combustible substances
Molecular Formula (NH4)2S2O8
Purity ACS, Reag. Ph Eur
Odor Odorless
Solubility Information Soluble in water: 620g/L at 20°C

Specifications

Formula Weight 228.19
CAS 7727-54-0
Chemical Name or Material Ammonium peroxodisulfate
Density 1.98g/cm3 at 20°C
Molecular Formula (NH4)2S2O8
Solubility Information Soluble in water: 620g/L at 20°C
Recommended Storage Store at 15° to 25°C (59° to 77°F), Tightly closed. Dry. Away from combustible substances
Synonym Ammonium persulfate; Peroxidisulfuric acid diammonium salt
Accessories

H272: May intensify fire; oxidiser.

H315: Causes skin irritation.

H302: Harmful if swallowed.

Store at 15° to 25°C (59° to 77°F), Tightly closed. Dry. Away from combustible substances