missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium perchlorate, Reagent Grade, Alfa Aesar

$187.80 - $187.80

Chemical Identifiers

CAS 7790-98-9
Physical Form Crystalline
Molecular Formula H4ClNO4
Formula Weight 117.5
MDL Number MFCD00011421
Chemical Name or Material Ammonium perchlorate
Grade Reagent Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1165830 Alfa Aesar
1165830
250g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA11658A1 Alfa Aesar
11658A1
1kg Each for $187.80
Specifications Description & Specifications

Chemical Identifiers

7790-98-9
H4ClNO4
MFCD00011421
Reagent Grade
Crystalline
117.5
Ammonium perchlorate

Specifications

117.5
White
Crystalline
Odorless
7790-98-9
Ammonium perchlorate
250g
1.95g/cm3
H4ClNO4
MFCD00011421
White crystalline
Reagent Grade

Safety and Handling

H200-H271-H302

DOTInformation : DOT Class: 5.1, Packaging Group: II

EINECSNumber : 232-235-1